siRNA Details
| Gene Symbol: Zfp397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271475.1 | NP_001258404.1 | 4 | 1047 | ||
| XM_006254456.2 | XP_006254518.1 | 4 | 1064 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp397| Gene Symbol: Zfp397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271475.1 | NP_001258404.1 | 4 | 1047 | ||
| XM_006254456.2 | XP_006254518.1 | 4 | 1064 | |||