siRNA Details
| Gene Symbol: Kmt2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235840.2 | XP_006235902.1 | 17 | 5482 | ||
| XM_008762670.1 | XP_008760892.1 | 56 | 14754 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kmt2c| Gene Symbol: Kmt2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235840.2 | XP_006235902.1 | 17 | 5482 | ||
| XM_008762670.1 | XP_008760892.1 | 56 | 14754 | |||