siRNA Details
| Gene Symbol: March9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100601.1 | NP_001094071.1 | 3 | 607 | ||
| XM_008765422.1 | XP_008763644.1 | 4 | 1582 | |||
| GenBank mRNA | AB048842.1 | - | 3 | 607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

March9| Gene Symbol: March9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100601.1 | NP_001094071.1 | 3 | 607 | ||
| XM_008765422.1 | XP_008763644.1 | 4 | 1582 | |||
| GenBank mRNA | AB048842.1 | - | 3 | 607 | ||