siRNA Details
| Gene Symbol: RGD1309870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251478.2 | XP_006251540.1 | 1 | 635 | ||
| XM_006251479.2 | XP_006251541.1 | 1 | 635 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309870| Gene Symbol: RGD1309870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251478.2 | XP_006251540.1 | 1 | 635 | ||
| XM_006251479.2 | XP_006251541.1 | 1 | 635 | |||