siRNA Details
| Gene Symbol: Sepsecs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128287.2 | NP_001121759.1 | 8, 9 | 1035 | ||
| GenBank mRNA | BC166584.1 | - | 8, 9 | 1035 | 7443474 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sepsecs| Gene Symbol: Sepsecs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128287.2 | NP_001121759.1 | 8, 9 | 1035 | ||
| GenBank mRNA | BC166584.1 | - | 8, 9 | 1035 | 7443474 | |