siRNA Details
| Gene Symbol: Tspan15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115032.1 | NP_001108504.1 | 5, 6 | 672 | ||
| GenBank mRNA | BC158654.1 | - | 5, 6 | 672 | 8368493 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspan15| Gene Symbol: Tspan15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115032.1 | NP_001108504.1 | 5, 6 | 672 | ||
| GenBank mRNA | BC158654.1 | - | 5, 6 | 672 | 8368493 | |