siRNA Details
| Gene Symbol: Vrtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750199.3 | XP_003750247.1 | 1 | 1209 | ||
| XM_008764891.1 | XP_008763113.1 | 1 | 1209 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vrtn| Gene Symbol: Vrtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750199.3 | XP_003750247.1 | 1 | 1209 | ||
| XM_008764891.1 | XP_008763113.1 | 1 | 1209 | |||