siRNA Details
| Gene Symbol: Cntln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114402.1 | NP_001107874.1 | 3, 4 | 736 | ||
| GenBank mRNA | AB369315.1 | - | 3, 4 | 736 | ||
| CB699924.1 | - | 1, 2 | 370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntln| Gene Symbol: Cntln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114402.1 | NP_001107874.1 | 3, 4 | 736 | ||
| GenBank mRNA | AB369315.1 | - | 3, 4 | 736 | ||
| CB699924.1 | - | 1, 2 | 370 | |||