siRNA Details
| Gene Symbol: Dram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173427.1 | NP_001166898.1 | 1 | 175 | ||
| GenBank mRNA | BC168648.1 | - | 1 | 175 | 7099292 | |
| FQ211820.1 | - | 1 | 303 | |||
| FQ223303.1 | - | 1 | 236 | |||
| FQ229264.1 | - | 1 | 236 | |||

Dram1
Genomic Map