siRNA Details
| Gene Symbol: Dram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173427.1 | NP_001166898.1 | 2 | 297 | ||
| GenBank mRNA | BC168648.1 | - | 2 | 297 | 7099292 | |
| FQ211820.1 | - | 2 | 425 | |||
| FQ229264.1 | - | 2 | 358 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dram1| Gene Symbol: Dram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173427.1 | NP_001166898.1 | 2 | 297 | ||
| GenBank mRNA | BC168648.1 | - | 2 | 297 | 7099292 | |
| FQ211820.1 | - | 2 | 425 | |||
| FQ229264.1 | - | 2 | 358 | |||