siRNA Details
| Gene Symbol: Wbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127502.1 | NP_001120974.1 | 3 | 465 | ||
| XM_006257281.2 | XP_006257343.1 | 3 | 504 | |||
| XM_008773438.1 | XP_008771660.1 | 3 | 485 | |||
| GenBank mRNA | BC166897.1 | - | 3 | 465 | 6921863 | |
| FQ215245.1 | - | 3 | 393 | |||
| FQ220910.1 | - | 3 | 447 | |||
| FQ232216.1 | - | 3 | 439 | |||

Wbp5
Genomic Map