siRNA Details
Gene Symbol: Mbnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111064.1 | NP_001104534.1 | 4 | 1069 | ||
GenBank mRNA | AA818999.1 | - | 2 | 399 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mbnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111064.1 | NP_001104534.1 | 4 | 1069 | ||
GenBank mRNA | AA818999.1 | - | 2 | 399 |