siRNA Details
| Gene Symbol: LOC680663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127377.1 | NP_001120849.1 | 5 | 1245 | ||
| XM_008773439.1 | XP_008771661.1 | 5 | 1327 | |||
| GenBank mRNA | BC166523.1 | - | 5 | 1245 | 7134576 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC680663| Gene Symbol: LOC680663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127377.1 | NP_001120849.1 | 5 | 1245 | ||
| XM_008773439.1 | XP_008771661.1 | 5 | 1327 | |||
| GenBank mRNA | BC166523.1 | - | 5 | 1245 | 7134576 | |