siRNA Details
| Gene Symbol: Obp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128139.2 | NP_001121611.1 | 1 | 22 | ||
| XM_008761608.1 | XP_008759830.1 | 1 | 1345 | |||
| GenBank mRNA | DQ537493.1 | - | 1 | 24 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Obp2a| Gene Symbol: Obp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128139.2 | NP_001121611.1 | 1 | 22 | ||
| XM_008761608.1 | XP_008759830.1 | 1 | 1345 | |||
| GenBank mRNA | DQ537493.1 | - | 1 | 24 | ||