siRNA Details
| Gene Symbol: Fam132b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001060107.4 | XP_001060107.2 | 7 | 1035 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam132b| Gene Symbol: Fam132b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001060107.4 | XP_001060107.2 | 7 | 1035 | ||