siRNA Details
| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101002.1 | NP_001094472.1 | 8, 9 | 1667 | ||
| GenBank mRNA | BC128774.1 | - | 8, 9 | 1667 | 7132745 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pou6f2| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101002.1 | NP_001094472.1 | 8, 9 | 1667 | ||
| GenBank mRNA | BC128774.1 | - | 8, 9 | 1667 | 7132745 | |