siRNA Details
| Gene Symbol: MGC94891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044277.1 | NP_001037742.2 | 9 | 1374 | ||
| GenBank mRNA | BC083819.1 | - | 9 | 1374 | 7111103 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC94891| Gene Symbol: MGC94891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044277.1 | NP_001037742.2 | 9 | 1374 | ||
| GenBank mRNA | BC083819.1 | - | 9 | 1374 | 7111103 | |