siRNA Details
| Gene Symbol: Tmem230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048043.1 | NP_001041508.1 | 4 | 490 | ||
| XM_006235070.2 | XP_006235132.1 | 5 | 588 | |||
| GenBank mRNA | BC105614.1 | - | 4 | 490 | 7384127 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem230| Gene Symbol: Tmem230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048043.1 | NP_001041508.1 | 4 | 490 | ||
| XM_006235070.2 | XP_006235132.1 | 5 | 588 | |||
| GenBank mRNA | BC105614.1 | - | 4 | 490 | 7384127 | |