siRNA Details
| Gene Symbol: LOC681383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109441.1 | NP_001102911.1 | 6, 7 | 770 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC681383| Gene Symbol: LOC681383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109441.1 | NP_001102911.1 | 6, 7 | 770 | ||