siRNA Details
| Gene Symbol: Rnf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109444.2 | NP_001102914.1 | 741 | |||
| GenBank mRNA | BC081881.1 | - | 741 | 7108181 | ||
| FQ224754.1 | - | 909 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf13| Gene Symbol: Rnf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109444.2 | NP_001102914.1 | 741 | |||
| GenBank mRNA | BC081881.1 | - | 741 | 7108181 | ||
| FQ224754.1 | - | 909 | ||||