siRNA Details
| Gene Symbol: Vbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229645.2 | XP_006229707.1 | 4, 5 | 569 | ||
| XM_006229646.2 | XP_006229708.1 | 4, 5 | 584 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vbp1| Gene Symbol: Vbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229645.2 | XP_006229707.1 | 4, 5 | 569 | ||
| XM_006229646.2 | XP_006229708.1 | 4, 5 | 584 | |||