siRNA Details
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109445.1 | NP_001102915.1 | 1, 2 | 143 | ||
GenBank mRNA | BC158833.1 | - | 1, 2 | 85 | 7381517 | |
FJ179410.1 | - | 20 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109445.1 | NP_001102915.1 | 1, 2 | 143 | ||
GenBank mRNA | BC158833.1 | - | 1, 2 | 85 | 7381517 | |
FJ179410.1 | - | 20 |