siRNA Details
| Gene Symbol: Mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085406.1 | NP_001078875.1 | 8, 9 | 966 | ||
| XM_008766773.1 | XP_008764995.1 | 9, 10 | 1254 | |||
| GenBank mRNA | BC101893.1 | - | 7, 8 | 794 | ||
| BC158552.1 | - | 8, 9 | 966 | 9098910 | ||

Mpnd
Genomic Map