siRNA Details
| Gene Symbol: Vipas39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101004.1 | NP_001094474.1 | 17 | 1287 | ||
| XM_006240400.2 | XP_006240462.1 | 18 | 1478 | |||
| GenBank mRNA | BC087099.1 | - | 17 | 1287 | 7192222 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vipas39| Gene Symbol: Vipas39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101004.1 | NP_001094474.1 | 17 | 1287 | ||
| XM_006240400.2 | XP_006240462.1 | 18 | 1478 | |||
| GenBank mRNA | BC087099.1 | - | 17 | 1287 | 7192222 | |