siRNA Details
| Gene Symbol: Ahsa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115034.1 | NP_001108506.1 | 9 | 1097 | ||
| GenBank mRNA | BC158698.1 | - | 9 | 1054 | 9027714 | |
| FQ223544.1 | - | 9 | 1081 | |||
| FQ232113.1 | - | 9 | 1060 | |||
| FQ233084.1 | - | 9 | 1059 | |||

Ahsa1
Genomic Map