siRNA Details
| Gene Symbol: Eif3i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115035.1 | NP_001108507.1 | 569 | |||
| GenBank mRNA | BC158748.1 | - | 537 | 9101450 | ||
| BC158821.1 | - | 565 | 7375325 | |||
| FQ209543.1 | - | 551 | ||||
| FQ213040.1 | - | 550 | ||||
| FQ220584.1 | - | 553 | ||||
| FQ220807.1 | - | 554 | ||||
| FQ227121.1 | - | 555 | ||||
| FQ230070.1 | - | 555 | ||||
| FQ231084.1 | - | 545 | ||||
| FQ231314.1 | - | 545 | ||||

Eif3i
Genomic Map