siRNA Details
| Gene Symbol: Eif3i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115035.1 | NP_001108507.1 | 692 | |||
| GenBank mRNA | BC158748.1 | - | 660 | 9101450 | ||
| BC158821.1 | - | 688 | 7375325 | |||
| FQ209543.1 | - | 674 | ||||
| FQ213040.1 | - | 673 | ||||
| FQ220584.1 | - | 676 | ||||
| FQ220807.1 | - | 677 | ||||
| FQ230070.1 | - | 678 | ||||
| FQ231084.1 | - | 668 | ||||
| FQ231314.1 | - | 668 | ||||

Eif3i
Genomic Map