siRNA Details
| Gene Symbol: Eif3i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115035.1 | NP_001108507.1 | 961 | |||
| GenBank mRNA | BC158748.1 | - | 929 | 9101450 | ||
| BC158821.1 | - | 957 | 7375325 | |||
| FQ209543.1 | - | 943 | ||||
| FQ213040.1 | - | 942 | ||||
| FQ220584.1 | - | 945 | ||||
| FQ220807.1 | - | 946 | ||||
| FQ227121.1 | - | 950 | ||||
| FQ230070.1 | - | 947 | ||||
| FQ231084.1 | - | 937 | ||||
| FQ231314.1 | - | 937 | ||||

Eif3i
Genomic Map