siRNA Details
| Gene Symbol: Ccdc28b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134689.1 | NP_001128161.1 | 2 | 103 | ||
| XM_008764171.1 | XP_008762393.1 | 2 | 2898 | |||
| GenBank mRNA | BC168155.1 | - | 1 | 480 | 8362701 | |
| CB711403.1 | - | 2 | 113 | |||

Ccdc28b
Genomic Map