siRNA Details
| Gene Symbol: Esco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126299.2 | NP_001119771.2 | 1 | 1413 | ||
| XM_006254412.2 | XP_006254474.1 | 3 | 2026 | |||
| XM_006254413.2 | XP_006254475.1 | 3 | 1836 | |||
| XM_006254414.2 | XP_006254476.1 | 3 | 2023 | |||
| XM_006254415.2 | XP_006254477.1 | 3 | 2189 | |||
| GenBank mRNA | BC166441.1 | - | 1 | 1413 | 7461161 | |

Esco1
Genomic Map