siRNA Details
| Gene Symbol: LOC102552143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763961.1 | XP_008762183.1 | 3 | 563 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102552143| Gene Symbol: LOC102552143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763961.1 | XP_008762183.1 | 3 | 563 | ||