siRNA Details
| Gene Symbol: Muc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760048.1 | XP_008758270.1 | 14 | 2408 | ||
| GenBank mRNA | U07615.1 | - | 14 | 1810 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Muc2| Gene Symbol: Muc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760048.1 | XP_008758270.1 | 14 | 2408 | ||
| GenBank mRNA | U07615.1 | - | 14 | 1810 | ||