siRNA Details
Gene Symbol: LOC680200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139491.1 | NP_001132963.1 | 4 | 1167 | ||
GenBank mRNA | BC167062.1 | - | 4 | 1167 | 9024521 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC680200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139491.1 | NP_001132963.1 | 4 | 1167 | ||
GenBank mRNA | BC167062.1 | - | 4 | 1167 | 9024521 |