siRNA Details
| Gene Symbol: Rcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128189.2 | NP_001121661.1 | 8 | 902 | ||
| XM_006239073.2 | XP_006239135.1 | 8 | 872 | |||
| XM_006239074.1 | XP_006239136.1 | 8 | 1196 | |||
| XM_006239075.2 | XP_006239137.1 | 8 | 1386 | |||
| XM_006239077.2 | XP_006239139.1 | 7 | 832 | |||
| XM_008764172.1 | XP_008762394.1 | 8 | 2540 | |||
| XM_008764173.1 | XP_008762395.1 | 9 | 926 | |||
| GenBank mRNA | BC160834.1 | - | 8 | 902 | 7456254 | |

Rcc1
Genomic Map