siRNA Details
| Gene Symbol: Ankhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204053.1 | NP_001190982.1 | 3 | 612 | ||
| GenBank mRNA | FQ086998.1 | - | 3 | 612 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankhd1| Gene Symbol: Ankhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204053.1 | NP_001190982.1 | 3 | 612 | ||
| GenBank mRNA | FQ086998.1 | - | 3 | 612 | ||