siRNA Details
| Gene Symbol: Isx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751833.3 | XP_003751881.1 | 2 | 245 | ||
| XM_008772332.1 | XP_008770554.1 | 2 | 245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isx| Gene Symbol: Isx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751833.3 | XP_003751881.1 | 2 | 245 | ||
| XM_008772332.1 | XP_008770554.1 | 2 | 245 | |||