siRNA Details
| Gene Symbol: Pja1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101006.1 | NP_001094476.1 | 3 | 796 | ||
| XM_006257105.2 | XP_006257167.1 | 2 | 1910 | |||
| XM_006257106.2 | XP_006257168.1 | 2 | 1627 | |||
| XM_006257107.2 | XP_006257169.1 | 3 | 1370 | |||
| GenBank mRNA | BC081885.1 | - | 3 | 796 | 7108525 | |

Pja1
Genomic Map