siRNA Details
| Gene Symbol: Mogat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109436.2 | NP_001102906.2 | 4 | 629 | ||
| GenBank mRNA | BC166995.1 | - | 4 | 629 | 9027036 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mogat2| Gene Symbol: Mogat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109436.2 | NP_001102906.2 | 4 | 629 | ||
| GenBank mRNA | BC166995.1 | - | 4 | 629 | 9027036 | |