siRNA Details
| Gene Symbol: Ap3s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115039.1 | NP_001108511.1 | 4 | 455 | ||
| GenBank mRNA | BC158881.1 | - | 4 | 455 | 7936771 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap3s2| Gene Symbol: Ap3s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115039.1 | NP_001108511.1 | 4 | 455 | ||
| GenBank mRNA | BC158881.1 | - | 4 | 455 | 7936771 | |