siRNA Details
| Gene Symbol: LOC102552318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760184.1 | XP_008758406.1 | 10 | 1503 | ||
| XM_008760185.1 | XP_008758407.1 | 9 | 1313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102552318| Gene Symbol: LOC102552318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760184.1 | XP_008758406.1 | 10 | 1503 | ||
| XM_008760185.1 | XP_008758407.1 | 9 | 1313 | |||