siRNA Details
| Gene Symbol: Zfp639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080907.2 | NP_001074376.1 | 6 | 1060 | ||
| XM_006232254.2 | XP_006232316.1 | 7 | 1130 | |||
| XM_008760942.1 | XP_008759164.1 | 6 | 2609 | |||
| XM_008760943.1 | XP_008759165.1 | 7 | 1881 | |||
| XM_008760944.1 | XP_008759166.1 | 4 | 1651 | |||
| GenBank mRNA | BC087704.1 | - | 6 | 1061 | 7313723 | |

Zfp639
Genomic Map