siRNA Details
| Gene Symbol: Gnpda1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134995.1 | NP_001128467.1 | 4, 5 | 809 | ||
| GenBank mRNA | BC169009.1 | - | 4, 5 | 771 | 7312423 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnpda1| Gene Symbol: Gnpda1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134995.1 | NP_001128467.1 | 4, 5 | 809 | ||
| GenBank mRNA | BC169009.1 | - | 4, 5 | 771 | 7312423 | |