siRNA Details
| Gene Symbol: NEWGENE_1585613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761114.1 | XP_008759336.1 | 1 | 543 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEWGENE_1585613| Gene Symbol: NEWGENE_1585613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761114.1 | XP_008759336.1 | 1 | 543 | ||