siRNA Details
| Gene Symbol: Sri | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128190.1 | NP_001121662.1 | 3 | 203 | ||
| XM_006236007.2 | XP_006236069.1 | 3 | 236 | |||
| GenBank mRNA | BC158845.1 | - | 3 | 203 | 7442862 | |
| FQ220011.1 | - | 3 | 215 | |||
| FQ226061.1 | - | 3 | 213 | |||
| FQ226813.1 | - | 3 | 213 | |||
| FQ227927.1 | - | 3 | 198 | |||
| FQ232170.1 | - | 3 | 213 | |||
| FQ234730.1 | - | 3 | 215 | |||

Sri
Genomic Map