siRNA Details
| Gene Symbol: LOC102551431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257108.2 | XP_006257170.2 | 1 | 226 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102551431| Gene Symbol: LOC102551431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257108.2 | XP_006257170.2 | 1 | 226 | ||