siRNA Details
| Gene Symbol: Gal3st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109447.1 | NP_001102917.1 | 4 | 1531 | ||
| XM_006251382.2 | XP_006251444.1 | 3 | 1504 | |||
| XM_006251383.2 | XP_006251445.1 | 3 | 3145 | |||
| XM_008770365.1 | XP_008768587.1 | 2 | 1226 | |||
| XM_008770366.1 | XP_008768588.1 | 3 | 1607 | |||
| GenBank mRNA | BC087145.1 | - | 3 | 1281 | 7129361 | |

Gal3st1
Genomic Map