siRNA Details
| Gene Symbol: Gal3st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109447.1 | NP_001102917.1 | 2, 3 | 309 | ||
| XM_008770366.1 | XP_008768588.1 | 1, 2 | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gal3st1| Gene Symbol: Gal3st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109447.1 | NP_001102917.1 | 2, 3 | 309 | ||
| XM_008770366.1 | XP_008768588.1 | 1, 2 | 385 | |||