siRNA Details
| Gene Symbol: Rasef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277334.1 | NP_001264263.1 | 12, 13 | 1635 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasef| Gene Symbol: Rasef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277334.1 | NP_001264263.1 | 12, 13 | 1635 | ||