siRNA Details
| Gene Symbol: LOC683722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101008.1 | NP_001094478.1 | 1 | 168 | ||
| XM_008758772.1 | XP_008756994.1 | 1 | 420 | |||
| XM_008758773.1 | XP_008756995.1 | 1 | 417 | |||
| XM_008758774.1 | XP_008756996.1 | 1 | 273 | |||
| GenBank mRNA | BC098026.1 | - | 1 | 168 | 7455942 | |

LOC683722
Genomic Map