siRNA Details
| Gene Symbol: Prr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728150.4 | XP_002728196.2 | 2 | 174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr32| Gene Symbol: Prr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728150.4 | XP_002728196.2 | 2 | 174 | ||